Alfa Aesar™ 4,5,6,7-Tetrachlorofluorescein

Manufacturer: Alfa Aesar™ B2563506

Quantity 5g

 View more versions of this product

Catalog No. AAB2563506

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 6262-21-1
    Molecular Formula C20H8Cl4O5
    Formula Weight 470.1
    MDL Number MFCD00010030
    Synonym 4,5,6,7-Tetrachloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]-xanthene]-3-one
    Chemical Name or Material 4,5,6,7-Tetrachlorofluorescein

    Specifications

    Formula Weight 470.1
    TSCA TSCA
    Molecular Formula C20H8Cl4O5
    MDL Number MFCD00010030
    CAS 6262-21-1
    Synonym 4,5,6,7-Tetrachloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]-xanthene]-3-one
    Melting Point 304°C (dec.)
    Quantity 5g
    Chemical Name or Material 4,5,6,7-Tetrachlorofluorescein

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit