Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
879668-17-4
C6H10ClN3
159.617
MFCD11846200
CHLFJSVTUQAGSC-UHFFFAOYSA-N
4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hydrochloride,4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl,4,5,6,7-tetrahydro-1h-imidazo 4,5-c pyridine hydrochloride,3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride,4,5,6,7-tetrahydro-5-azabenzimidazole hydrochloride,3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloridyl,4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl,1h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride,4,5,6,7-tetrahydro-1h-imidazo 3,4-c pyridine hydrochloride,4,5,6,7-tetrahydro-imidazo-4,5-c-pyridine hydrochloride
21330723
4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine;hydrochloride
C1CNCC2=C1N=CN2.Cl
879668-17-4
C6H10ClN3
159.617
MFCD11846200
CHLFJSVTUQAGSC-UHFFFAOYSA-N
4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hydrochloride,4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl,4,5,6,7-tetrahydro-1h-imidazo 4,5-c pyridine hydrochloride,3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride,4,5,6,7-tetrahydro-5-azabenzimidazole hydrochloride,3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloridyl,4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl,1h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride,4,5,6,7-tetrahydro-1h-imidazo 3,4-c pyridine hydrochloride,4,5,6,7-tetrahydro-imidazo-4,5-c-pyridine hydrochloride
21330723
4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine;hydrochloride
C1CNCC2=C1N=CN2.Cl
Quantity 1g
Beilstein 15312806
Formula Weight 159.62
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride