Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
40133-07-1
C9H10O2S
182.237
ROLXOQXKNDKXTA-UHFFFAOYSA-N
4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylic acid,4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic acid,benzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro,acmc-1asbb,enamine_001826,2-carboxy-4,5,6,7-tetrahydro-1-benzothiophene,2-carboxy-4,5,6,7-tetrahydrobenzo b thiophene,4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylicacid,4,5,6,7-tetrahydrobenzo b-thiophene-2-carboxylic acid,4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic
2063443
4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylic acid
C1CCC2=C(C1)C=C(S2)C(=O)O
40133-07-1
C9H10O2S
182.237
ROLXOQXKNDKXTA-UHFFFAOYSA-N
4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylic acid,4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic acid,benzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro,acmc-1asbb,enamine_001826,2-carboxy-4,5,6,7-tetrahydro-1-benzothiophene,2-carboxy-4,5,6,7-tetrahydrobenzo b thiophene,4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylicacid,4,5,6,7-tetrahydrobenzo b-thiophene-2-carboxylic acid,4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic
2063443
4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylic acid
C1CCC2=C(C1)C=C(S2)C(=O)O
Quantity 1g
Color White
Melting Point 195°C to 205°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 182.24
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.