Alfa Aesar™ 4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene, 99%

Manufacturer: Alfa Aesar™ H2742006

Quantity 5g

 View more versions of this product

Catalog No. AAH2742006

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C31H48OP2
    Formula Weight 498.68
    MDL Number MFCD07781995
    Synonym t-Bu-Xantphos
    Chemical Name or Material 4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene

    Specifications

    Formula Weight 498.68
    Molecular Formula C31H48OP2
    MDL Number MFCD07781995
    Synonym t-Bu-Xantphos
    Melting Point 153° to 157°C
    Quantity 5g
    Chemical Name or Material 4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit