Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4',5'-Dibromofluorescein, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
596-03-2
C20H10Br2O5
490.11
ZDTNHRWWURISAA-UHFFFAOYSA-N
4',5'-dibromofluorescein,solvent red 72,eosinic acid,japan orange 201,FD&C orange no. 5,c.i. solvent red 72,4',5'-dibromofluororescein,4,5-dibromofluorescein,d & c orange no. 5,japan orange no. 201
11689
4',5'-dibromo-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)Br)OC5=C3C=CC(=C5Br)O
596-03-2
C20H10Br2O5
490.11
ZDTNHRWWURISAA-UHFFFAOYSA-N
4',5'-dibromofluorescein,solvent red 72,eosinic acid,japan orange 201,FD&C orange no. 5,c.i. solvent red 72,4',5'-dibromofluororescein,4,5-dibromofluorescein,d & c orange no. 5,japan orange no. 201
11689
4',5'-dibromo-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)Br)OC5=C3C=CC(=C5Br)O
Packaging Glass bottle
Melting Point 270.0°C to 273.0°C
Quantity 5g
Loss on Drying 5% max.
Infrared Spectrum Authentic
Beilstein 19,V,6,462
Merck Index 15,3027
Solubility Information (1% in hot 0.5 N NH4OH) Clear yellow to orange
Formula Weight 490.11
Chemical Name or Material 4',5'-Dibromofluorescein

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 209-876-