Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dichloro-2-(4-methylphenyl)-2,3-dihydropyridazin-3-one, 97%, Maybridge™

Click to view available options
Quantity:
1g
10g
33098-21-4
C11H8Cl2N2O
255.098
MFCD00135290
OWPCMQKRJFNLHF-UHFFFAOYSA-N
4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3-one, 4,5-dichloro-2-4-methylphenyl pyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-4-methylphenyl, 4,5-dichloro-2-p-tolyl-2h-pyridazin-3-one, 4,5-dichloro-2-4-methylphenyl-2-hydropyridazin-3-one, 4,5-dichloro-2-p-tolyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-3 2h-pyridazinone, 4,5-dichloro-2-4-methylphenyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3one
2774757
4,5-dichloro-2-(4-methylphenyl)pyridazin-3-one
CC1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Cl)Cl
33098-21-4
C11H8Cl2N2O
255.098
MFCD00135290
OWPCMQKRJFNLHF-UHFFFAOYSA-N
4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3-one, 4,5-dichloro-2-4-methylphenyl pyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-4-methylphenyl, 4,5-dichloro-2-p-tolyl-2h-pyridazin-3-one, 4,5-dichloro-2-4-methylphenyl-2-hydropyridazin-3-one, 4,5-dichloro-2-p-tolyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-3 2h-pyridazinone, 4,5-dichloro-2-4-methylphenyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3one
2774757
4,5-dichloro-2-(4-methylphenyl)pyridazin-3-one
CC1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Cl)Cl
Quantity 1g
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 255.1
Percent Purity 97%
Chemical Name or Material 4, 5-Dichloro-2-(4-methylphenyl)-2, 3-dihydropyridazin-3-one

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.