Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dichlorophthalic anhydride, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
942-06-3
C8H2Cl2O3
217.01
MFCD00075034
ULSOWUBMELTORB-UHFFFAOYSA-N
4,5-dichlorophthalic anhydride,5,6-dichloroisobenzofuran-1,3-dione,4,5-dichlorophthalicanhydride,1,3-isobenzofurandione, 5,6-dichloro,5,6-dichloro-1,3-dihydro-2-benzofuran-1,3-dione,5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione,5,6-dichlor-2-benzofuran-1,3-dion,pubchem16761,acmc-209rqk,4,5-dichlorophalic anhydride
70334
5,6-dichloro-2-benzofuran-1,3-dione
C1=C2C(=CC(=C1Cl)Cl)C(=O)OC2=O
942-06-3
C8H2Cl2O3
217.01
MFCD00075034
ULSOWUBMELTORB-UHFFFAOYSA-N
4,5-dichlorophthalic anhydride,5,6-dichloroisobenzofuran-1,3-dione,4,5-dichlorophthalicanhydride,1,3-isobenzofurandione, 5,6-dichloro,5,6-dichloro-1,3-dihydro-2-benzofuran-1,3-dione,5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione,5,6-dichlor-2-benzofuran-1,3-dion,pubchem16761,acmc-209rqk,4,5-dichlorophalic anhydride
70334
5,6-dichloro-2-benzofuran-1,3-dione
C1=C2C(=CC(=C1Cl)Cl)C(=O)OC2=O
Melting Point 185°C to 187°C
Quantity 1g
Color Blue to Gray
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 217.01
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4, 5-Dichlorophthalic anhydride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-386-2