Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Difluoro-2-nitroaniline, 98%, Thermo Scientific™

Catalog Number ac397020010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
78056-39-0
C6H4F2N2O2
174.11
MFCD00010876
WDMCABATCGQAKK-UHFFFAOYSA-N
3,4-difluoro-6-nitroaniline, benzenamine, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitro-aniline, 4,5-difluoro-2-nitrophenylamine, 2-nitro-4,5-difluoroaniline, 2-amino-4,5-difluoronitrobenzene, 4,5-difluoro-2-nitro-phenylamine, 4,5-difluoro-2-nitrobenzenamine, pubchem2121, acmc-209pdb
157151
4,5-difluoro-2-nitroaniline
NC1=CC(F)=C(F)C=C1[N+]([O-])=O
78056-39-0
C6H4F2N2O2
174.11
MFCD00010876
WDMCABATCGQAKK-UHFFFAOYSA-N
3,4-difluoro-6-nitroaniline, benzenamine, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitro-aniline, 4,5-difluoro-2-nitrophenylamine, 2-nitro-4,5-difluoroaniline, 2-amino-4,5-difluoronitrobenzene, 4,5-difluoro-2-nitro-phenylamine, 4,5-difluoro-2-nitrobenzenamine, pubchem2121, acmc-209pdb
157151
4,5-difluoro-2-nitroaniline
NC1=CC(F)=C(F)C=C1[N+]([O-])=O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 107°C to 109°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H2(NO2)NH2
Quantity 1 g
Formula Weight 174.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 5-Difluoro-2-nitroaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit