Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Croconic acid, 97%, Acros Organics™

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
488-86-8
C5H2O5
142.066
MFCD00181389
RBSLJAJQOVYTRQ-UHFFFAOYSA-N
croconic acid,crocic acid,4,5-dihydroxy-4-cyclopentene-1,2,3-trione,unii-6hgy35uxaa,6hgy35uxaa,4-cyclopentene-1,2,3-trione, 4,5-dihydroxy,croconicacid,4,5-dihydroxy-4-cyclopentene-1,2,3-trione #
546874
4,5-dihydroxycyclopent-4-ene-1,2,3-trione
C1(=C(C(=O)C(=O)C1=O)O)O
488-86-8
C5H2O5
142.066
MFCD00181389
RBSLJAJQOVYTRQ-UHFFFAOYSA-N
croconic acid,crocic acid,4,5-dihydroxy-4-cyclopentene-1,2,3-trione,unii-6hgy35uxaa,6hgy35uxaa,4-cyclopentene-1,2,3-trione, 4,5-dihydroxy,croconicacid,4,5-dihydroxy-4-cyclopentene-1,2,3-trione #
546874
4,5-dihydroxycyclopent-4-ene-1,2,3-trione
C1(=C(C(=O)C(=O)C1=O)O)O
Packaging Glass bottle
Color Yellow
Quantity 500mg
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 142.07
Physical Form Powder or Crystals
Percent Purity ≥96.0%
Chemical Name or Material 4,5-Dihydroxy-4-cyclopentene-1,2,3-trione
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning