Alfa Aesar™ 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, ACS

Manufacturer: Alfa Aesar™ 2241406

Quantity 5g

 View more versions of this product

Catalog No. AA2241406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5808-22-0
    Formula Weight 400.29 (364.26 Anhydrous)
    MDL Number MFCD00150612
    Beilstein 3647189
    Merck Index 14,2241
    Synonym Chromotropic acid disodium salt dihydrate
    Chemical Name or Material 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate
    Grade ACS Grade

    Specifications

    Formula Weight 400.29 (364.26 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Off-White to Buff-Colored
    MDL Number MFCD00150612
    Health Hazard 1 Warning
    Melting Point >300°C
    Physical Form Powder
    CAS 5808-22-0
    Merck Index 14,2241
    Synonym Chromotropic acid disodium salt dihydrate
    Quantity 5g
    TSCA TSCA
    Beilstein 3647189
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate
    Grade ACS Grade

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit