Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethoxy-2-nitrobenzoic acid, 99%, Acros Organics™

Click to view available options
Quantity:
5g
100g
4998-07-6
C9H9NO6
227.17
WWCMFGBGMJAJRX-UHFFFAOYSA-M
4,5-dimethoxy-2-nitrobenzoic acid,6-nitroveratric acid,3,4-dimethoxy-6-nitrobenzoic acid,benzoic acid, 4,5-dimethoxy-2-nitro,zlchem 681,acmc-2097gn,ksc496i5r,4,5-dimethoxy-2-nitrobenzoicacid,2-nitro-4,5-dimethoxybenzoic acid,4,5-dimethoxy-2-nitro-benzoic acid
78690
COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC
4998-07-6
C9H9NO6
227.17
WWCMFGBGMJAJRX-UHFFFAOYSA-M
4,5-dimethoxy-2-nitrobenzoic acid,6-nitroveratric acid,3,4-dimethoxy-6-nitrobenzoic acid,benzoic acid, 4,5-dimethoxy-2-nitro,zlchem 681,acmc-2097gn,ksc496i5r,4,5-dimethoxy-2-nitrobenzoicacid,2-nitro-4,5-dimethoxybenzoic acid,4,5-dimethoxy-2-nitro-benzoic acid
78690
COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC
Packaging Glass Bottle
Quantity 5g
Melting Point 194.0°C to 195.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 227.17
Percent Purity 99%
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzoic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 225-657-2