Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethyl-2-furancarboxaldehyde, 97%, Acros Organics

Click to view available options
Quantity:
1g
52480-43-0
C7H8O2
124.139
JPTPEPVCVXGNJM-UHFFFAOYSA-N
4,5-dimethyl-2-furaldehyde,4,5-dimethylfurfural,4,5-dimethyl-furan-2-carbaldehyde,2-furancarboxaldehyde, 4,5-dimethyl,2-furanmethanethiolformate,acmc-1ao31,4.5-dimethyl-2-furaldehyde,4,5-dimethyl-2-furancarbaldehyde,4,5-dimethyl-2-furancarboxaldehyde,2-furancarboxaldehyde,4,5-dimethyl
643406
4,5-dimethylfuran-2-carbaldehyde
CC1=C(OC(=C1)C=O)C
52480-43-0
C7H8O2
124.139
JPTPEPVCVXGNJM-UHFFFAOYSA-N
4,5-dimethyl-2-furaldehyde,4,5-dimethylfurfural,4,5-dimethyl-furan-2-carbaldehyde,2-furancarboxaldehyde, 4,5-dimethyl,2-furanmethanethiolformate,acmc-1ao31,4.5-dimethyl-2-furaldehyde,4,5-dimethyl-2-furancarbaldehyde,4,5-dimethyl-2-furancarboxaldehyde,2-furancarboxaldehyde,4,5-dimethyl
643406
4,5-dimethylfuran-2-carbaldehyde
CC1=C(OC(=C1)C=O)C
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 1g
Refractive Index 1.5290 to 1.5320 (20°C, 589nm)
Density 1.01g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 124.14
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 4, 5-Dimethyl-2-furancarboxaldehyde, 97%
Show More Show Less

Recommended Storage : Product darkens on storage