Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethyl-2-nitroaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
6972-71-0
C8H10N2O2
166.18
MFCD00007811
PINGKGKKUSYUAW-UHFFFAOYSA-N
6-nitro-3,4-xylidine,benzenamine, 4,5-dimethyl-2-nitro,3,4-xylidine, 6-nitro,2-nitro-4,5-dimethylaniline,4,5-dimethyl-2-nitrophenylamine,2-nitro-4,5-xylidine,3,4-dimethyl-6-nitroaniline,acmc-20amrx,3, 6-nitro,4-amino-5-nitro-o-xylene
81445
4,5-dimethyl-2-nitroaniline
CC1=CC(=C(C=C1C)[N+](=O)[O-])N
6972-71-0
C8H10N2O2
166.18
MFCD00007811
PINGKGKKUSYUAW-UHFFFAOYSA-N
6-nitro-3,4-xylidine,benzenamine, 4,5-dimethyl-2-nitro,3,4-xylidine, 6-nitro,2-nitro-4,5-dimethylaniline,4,5-dimethyl-2-nitrophenylamine,2-nitro-4,5-xylidine,3,4-dimethyl-6-nitroaniline,acmc-20amrx,3, 6-nitro,4-amino-5-nitro-o-xylene
81445
4,5-dimethyl-2-nitroaniline
CC1=CC(=C(C=C1C)[N+](=O)[O-])N
Melting Point 138°C to 143°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Color Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)2C6H2(NO2)NH2
UN Number 2811
Beilstein 12, 1106
Formula Weight 166.18
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 4, 5-Dimethyl-2-nitroaniline
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning