Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethyl-o-phenylenediamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3171-45-7
C8H12N2
136.19
XSZYBMMYQCYIPC-UHFFFAOYSA-N
4,5-dimethyl-1,2-phenylenediamine,4,5-diamino-o-xylene,4,5-dimethyl-o-phenylenediamine,1,2-benzenediamine, 4,5-dimethyl,1,2-diamino-4,5-dimethylbenzene,4,5-dimethyl-1,2-benzenediamine,4,5-dimethyl-benzene-1,2-diamine,4,5-dimethyl-ortho-phenylenediamine,4,5-dimethylphenylene-1,2-diamine,4,5-dimethyl-o-phenylenediamine monohydrate
76635
4,5-dimethylbenzene-1,2-diamine
CC1=CC(=C(C=C1C)N)N
3171-45-7
C8H12N2
136.19
XSZYBMMYQCYIPC-UHFFFAOYSA-N
4,5-dimethyl-1,2-phenylenediamine,4,5-diamino-o-xylene,4,5-dimethyl-o-phenylenediamine,1,2-benzenediamine, 4,5-dimethyl,1,2-diamino-4,5-dimethylbenzene,4,5-dimethyl-1,2-benzenediamine,4,5-dimethyl-benzene-1,2-diamine,4,5-dimethyl-ortho-phenylenediamine,4,5-dimethylphenylene-1,2-diamine,4,5-dimethyl-o-phenylenediamine monohydrate
76635
4,5-dimethylbenzene-1,2-diamine
CC1=CC(=C(C=C1C)N)N
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Melting Point 127.2°C to 128.3°C
Color Brown to White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2C6H2(NH2)2
Beilstein 13,179
Solubility Information Solubility in water: slightly soluble. Other solubilities: good soluble in acidified water,soluble in alcohols and polar solvents
Formula Weight 136.19
Physical Form Crystalline Mass, Crystals or Chunks
Percent Purity 98%
Chemical Name or Material 4, 5-Dimethyl-o-phenylenediamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning