Alfa Aesar™ 4,5-Diphenyl-1,2,3-thiadiazole, 97%

Manufacturer: Alfa Aesar™ B2514006

Quantity 5g

 View more versions of this product

Catalog No. AAB2514006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5393-99-7
    Molecular Formula C14H10N2S
    Formula Weight 238.31
    MDL Number MFCD00082687
    Chemical Name or Material 4,5-Diphenyl-1,2,3-thiadiazole

    Specifications

    Formula Weight 238.31
    Molecular Formula C14H10N2S
    MDL Number MFCD00082687
    CAS 5393-99-7
    Melting Point 92° to 94°C
    Quantity 5g
    Chemical Name or Material 4,5-Diphenyl-1,2,3-thiadiazole
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit