Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Diphenylimidazole 98%, Acros Organics

Click to view available options
Quantity:
25g
668-94-0
C15H12N2
220.275
MFCD00005198
CPHGOBGXZQKCKI-UHFFFAOYSA-N
4,5-diphenylimidazole,diphenylimidazole,1h-imidazole, 4,5-diphenyl,imidazole, 4,5-diphenyl,dsstox_cid_25700,dsstox_rid_81072,dsstox_gsid_45700,4,5-diphenyl-1h-imidazol,imidazole,5-diphenyl,4,5-diphenylglyoxaline
69588
4,5-diphenyl-1H-imidazole
C1=CC=C(C=C1)C2=C(N=CN2)C3=CC=CC=C3
668-94-0
C15H12N2
220.275
MFCD00005198
CPHGOBGXZQKCKI-UHFFFAOYSA-N
4,5-diphenylimidazole,diphenylimidazole,1h-imidazole, 4,5-diphenyl,imidazole, 4,5-diphenyl,dsstox_cid_25700,dsstox_rid_81072,dsstox_gsid_45700,4,5-diphenyl-1h-imidazol,imidazole,5-diphenyl,4,5-diphenylglyoxaline
69588
4,5-diphenyl-1H-imidazole
C1=CC=C(C=C1)C2=C(N=CN2)C3=CC=CC=C3
Color Beige
Quantity 25g
Melting Point 229°C to 233°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 23, 255
Formula Weight 220.27
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4, 5-Diphenylimidazole, 98%
Show More Show Less

EINECSNumber : 211-573-3