Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,6-Dibromodibenzothiophene, 96%

Click to view available options
Quantity:
250mg
1g
669773-34-6
C12H6Br2S
342.048
MFCD18451473
ULGFJZPCGNTWFK-UHFFFAOYSA-N
4,6-dibromodibenzo b,d thiophene,4,6-dibromo-dibenzothiophene,6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene,dibenzothiophene, 4,6-dibromo,6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene
5245996
4,6-dibromodibenzothiophene
C1=CC2=C(C(=C1)Br)SC3=C2C=CC=C3Br
669773-34-6
C12H6Br2S
342.048
MFCD18451473
ULGFJZPCGNTWFK-UHFFFAOYSA-N
4,6-dibromodibenzo b,d thiophene,4,6-dibromo-dibenzothiophene,6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene,dibenzothiophene, 4,6-dibromo,6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene
5245996
4,6-dibromodibenzothiophene
C1=CC2=C(C(=C1)Br)SC3=C2C=CC=C3Br
Quantity 250mg
Color White
Formula Weight 342.05
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 4,6-Dibromodibenzothiophene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning