Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
705-24-8
C5HCl2F3N2
216.98
QFWVAJQVYBRTCL-UHFFFAOYSA-N
4,6-dichloro-2-trifluoromethyl pyrimidine,4,6-dichloro-2-trifluoromethyl-pyrimidine,pyrimidine, 4,6-dichloro-2-trifluoromethyl,pubchem19441,ksc496e6h,4 pound not6-dichloro-2-trifluoromethylpyrimidine
15713197
4,6-dichloro-2-(trifluoromethyl)pyrimidine
C1=C(N=C(N=C1Cl)C(F)(F)F)Cl
705-24-8
C5HCl2F3N2
216.98
QFWVAJQVYBRTCL-UHFFFAOYSA-N
4,6-dichloro-2-trifluoromethyl pyrimidine,4,6-dichloro-2-trifluoromethyl-pyrimidine,pyrimidine, 4,6-dichloro-2-trifluoromethyl,pubchem19441,ksc496e6h,4 pound not6-dichloro-2-trifluoromethylpyrimidine
15713197
4,6-dichloro-2-(trifluoromethyl)pyrimidine
C1=C(N=C(N=C1Cl)C(F)(F)F)Cl
Boiling Point 50°C to 52°C (5.3 mbar)
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 216.98
Percent Purity 97%
Chemical Name or Material 4, 6-Dichloro-2-(trifluoromethyl)pyrimidine

GHS Signal Word: Warning