Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,6-Difluoroindole, 97%

Manufacturer:  Alfa Aesar™ H54536MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH54536MD


  Check Availability
Add to cart

Specifications

Description

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Sparingly Soluble (0.062 g/L) (25°C), Calc.

Notes
Incompatible with oxidizing agents. Keep at room temperature.
Description & Specifications

Chemical Identifiers

199526-97-1
153.13
4,6-Difluoroindole
C8H5F2N
MFCD01075213

Specifications

250mg
C8H5F2N
MFCD01075213
97%
199526-97-1
153.13
4,6-Difluoroindole

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit