Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dihydroxy-2-methylmercaptopyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1979-98-2
C5H6N2O2S
158.17
AEXCUJUYEZIWJV-UHFFFAOYSA-N
4,6-dihydroxy-2-methylthiopyrimidine,2-methylthio pyrimidine-4,6-diol,s-methylthiobarbituric acid,4,6-dihydroxy-2-methylmercaptopyrimidine,2-methylthiopyrimidine-4,6-diol,4,6-dihydroxy-2-methythiopyrimidine,2-methylsulfanyl pyrimidine-4,6-diol,4,6-dihydroxy-2-methylthio pyrimidine,2-methylthio-4,6-pyrimidinediol
74795
4-hydroxy-2-methylsulfanyl-1H-pyrimidin-6-one
CSC1=NC(=CC(=O)N1)O
1979-98-2
C5H6N2O2S
158.17
AEXCUJUYEZIWJV-UHFFFAOYSA-N
4,6-dihydroxy-2-methylthiopyrimidine,2-methylthio pyrimidine-4,6-diol,s-methylthiobarbituric acid,4,6-dihydroxy-2-methylmercaptopyrimidine,2-methylthiopyrimidine-4,6-diol,4,6-dihydroxy-2-methythiopyrimidine,2-methylsulfanyl pyrimidine-4,6-diol,4,6-dihydroxy-2-methylthio pyrimidine,2-methylthio-4,6-pyrimidinediol
74795
4-hydroxy-2-methylsulfanyl-1H-pyrimidin-6-one
CSC1=NC(=CC(=O)N1)O
Packaging Glass Bottle
Color White
Melting Point >300.0°C
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 97%
Beilstein 25,63
Formula Weight 158.17
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4, 6-Dihydroxy-2-methylmercaptopyrimidine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.