Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2465-59-0
C5H4N4O2
152.11
MFCD00056934
HXNFUBHNUDHIGC-UHFFFAOYSA-N
oxypurinol,oxipurinol,alloxanthine,1h-pyrazolo 3,4-d pyrimidine-4,6-diol,oxoallopurinol,4,6-dihydroxypyrazolo 3,4-d pyrimidine,1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione,oxipurinolum,1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione,alloxanthin van
4644
CHEBI:28315
1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione
C1=C2C(=NC(=O)NC2=O)NN1
2465-59-0
C5H4N4O2
152.11
MFCD00056934
HXNFUBHNUDHIGC-UHFFFAOYSA-N
oxypurinol,oxipurinol,alloxanthine,1h-pyrazolo 3,4-d pyrimidine-4,6-diol,oxoallopurinol,4,6-dihydroxypyrazolo 3,4-d pyrimidine,1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione,oxipurinolum,1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione,alloxanthin van
4644
CHEBI:28315
1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione
C1=C2C(=NC(=O)NC2=O)NN1
Melting Point >300.0°C
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 152.11
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 4,6-Dihydroxypyrazolo[3,4-d]pyrimidine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Danger

EINECSNumber : 219-570-9