Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2465-59-0
C5H4N4O2
152.11
MFCD00056934
HXNFUBHNUDHIGC-UHFFFAOYSA-N
oxypurinol,oxipurinol,alloxanthine,1h-pyrazolo 3,4-d pyrimidine-4,6-diol,oxoallopurinol,4,6-dihydroxypyrazolo 3,4-d pyrimidine,1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione,oxipurinolum,1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione,alloxanthin van
4644
CHEBI:28315
1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione
C1=C2C(=NC(=O)NC2=O)NN1
2465-59-0
C5H4N4O2
152.11
MFCD00056934
HXNFUBHNUDHIGC-UHFFFAOYSA-N
oxypurinol,oxipurinol,alloxanthine,1h-pyrazolo 3,4-d pyrimidine-4,6-diol,oxoallopurinol,4,6-dihydroxypyrazolo 3,4-d pyrimidine,1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione,oxipurinolum,1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione,alloxanthin van
4644
CHEBI:28315
1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione
C1=C2C(=NC(=O)NC2=O)NN1
Infrared Spectrum Authentic
Melting Point >300.0°C
Quantity 25g
Packaging Glass bottle
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 152.11
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 4,6-Dihydroxypyrazolo[3,4-d]pyrimidine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning