Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dihydroxypyrimidine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
Plastic bottle
1193-24-4
C4H4N2O2
112.088
MFCD00016733
DUFGYCAXVIUXIP-UHFFFAOYSA-N
4,6-dihydroxypyrimidine,pyrimidine-4,6-diol,4,6-pyrimidinediol,6-hydroxy-4 1h-pyrimidinone,4 1h-pyrimidinone, 6-hydroxy,4 3h-pyrimidinone, 6-hydroxy,6-hydroxypyrimidin-4 3h-one,6-hydroxy-1h-pyrimidin-4-one,4,6-dihydroxypyrimidin,6-hydroxy-4-pyrimidinone
14512
4-hydroxy-1H-pyrimidin-6-one
C1=C(N=CNC1=O)O
1193-24-4
C4H4N2O2
112.088
MFCD00016733
DUFGYCAXVIUXIP-UHFFFAOYSA-N
4,6-dihydroxypyrimidine,pyrimidine-4,6-diol,4,6-pyrimidinediol,6-hydroxy-4 1h-pyrimidinone,4 1h-pyrimidinone, 6-hydroxy,4 3h-pyrimidinone, 6-hydroxy,6-hydroxypyrimidin-4 3h-one,6-hydroxy-1h-pyrimidin-4-one,4,6-dihydroxypyrimidin,6-hydroxy-4-pyrimidinone
14512
4-hydroxy-1H-pyrimidin-6-one
C1=C(N=CNC1=O)O
Color Yellow to Orange
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 112.09
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 4, 6-Dihydroxypyrimidine
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.