Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,6-Dimethylpyrimidine, 94%

Manufacturer:  Alfa Aesar™ A1030203

Quantity 1g

 View more versions of this product

Catalog No. AAA1030203


  Check Availability
Add to cart

Specifications

Description

Applications
4,6-Dimethylpyrimidine is used to produce (6-methyl-pyrimidin-4-yl)-pyruvic acid ethyl ester. It is used as pharmaceutical intermediate. It is also used in agrochemical and dyestuff field.

Solubility
Soluble in water at 20°C.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Description & Specifications

Chemical Identifiers

1558-17-4
108.14
UN1993
4,6-Dimethylpyrimidine
C6H8N2
MFCD00006116
107068

Specifications

46°C (114°F)
153° to 155°C
1558-17-4
108.14
UN1993
4,6-Dimethylpyrimidine
Recognizable
1g
C6H8N2
MFCD00006116
107068
94%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit