Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(6-Methyl-2-benzimidazolyl)benzamidoxime, 97%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
5g
1256486-22-2
C15H14N4O
266.304
MFCD16658868
SQIGGAWUCHCZKN-UHFFFAOYSA-N
4-6-methyl-2-benzimidazolyl benzamidoxime, afqxausvhhhsmn-uhfffaoysa-n, n-hydroxy-4-6-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide, n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide
73995333
[4-(5-methyl-1,3-dihydrobenzimidazol-2-ylidene)cyclohexa-2,5-dien-1-ylidene]-nitrosomethanamine
CC1=CC2=C(C=C1)NC(=C3C=CC(=C(N)N=O)C=C3)N2
1256486-22-2
C15H14N4O
266.304
MFCD16658868
SQIGGAWUCHCZKN-UHFFFAOYSA-N
4-6-methyl-2-benzimidazolyl benzamidoxime, afqxausvhhhsmn-uhfffaoysa-n, n-hydroxy-4-6-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide, n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide
73995333
[4-(5-methyl-1,3-dihydrobenzimidazol-2-ylidene)cyclohexa-2,5-dien-1-ylidene]-nitrosomethanamine
CC1=CC2=C(C=C1)NC(=C3C=CC(=C(N)N=O)C=C3)N2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Quantity 250mg
Formula Weight 266.3
Percent Purity 97%
Chemical Name or Material 4-(6-Methyl-2-benzimidazolyl)benzamidoxime

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures