Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-O-(4-Methoxybenzylidene)-D-glucal, 97%, Thermo Scientific™

Catalog Number aah60633md
Click to view available options
Quantity:
250 mg
1 g
312623-79-3
C14H16O5
264.277
MFCD01863528
SSKZQXMNNREVNP-HABKJSAYSA-N
4,6-o-4-methoxybenzylidene-d-glucal, 4 6-o-4-methoxybenzylidene-d-glucal, 4-o,6-o-4-methoxybenzylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,5-anhydro-4-o,6-o-4-methoxybenzylidene-2-deoxy-d-arabino-hexa-1-enitol, 4ar,8r,8as-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol
46185760
(4aR,8R,8aS)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
COC1=CC=C(C=C1)C2OCC3C(O2)C(C=CO3)O
312623-79-3
C14H16O5
264.277
MFCD01863528
SSKZQXMNNREVNP-HABKJSAYSA-N
4,6-o-4-methoxybenzylidene-d-glucal, 4 6-o-4-methoxybenzylidene-d-glucal, 4-o,6-o-4-methoxybenzylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,5-anhydro-4-o,6-o-4-methoxybenzylidene-2-deoxy-d-arabino-hexa-1-enitol, 4ar,8r,8as-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol
46185760
(4aR,8R,8aS)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
COC1=CC=C(C=C1)C2OCC3C(O2)C(C=CO3)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4,6-O-(4-Methoxybenzylidene)-D-glucal is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 264.27
Percent Purity 97%
Chemical Name or Material 4,6-O-(4-Methoxybenzylidene)-D-glucal
TSCA No
Recommended Storage Keep cold
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit