Learn More
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane, 97%, Thermo Scientific Chemicals
Description
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane is used as a phase transfer catalysts by transferring ions from one phase to another. It is involved in the synthesis of alkalides and electrides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Density | 1.11 g/mL |
Refractive Index | 1.501 |
Quantity | 0.1 g |
Beilstein | 616444 |
Sensitivity | Light sensitive |
Solubility Information | Miscible with water. |
Formula Weight | 332.44 |
Percent Purity | 97% |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 250-592-1 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.