Alfa Aesar™ 4,7-Dihydroxy-1,10-phenanthroline

Manufacturer: Alfa Aesar™ 3143402

Quantity 0.5g

 View more versions of this product

Catalog No. AA3143402

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3922-40-5
    Formula Weight 212.21
    MDL Number MFCD00004975
    Synonym 1,10-Phenanthroline-4,7-diol
    Chemical Name or Material 4,7-Dihydroxy-1,10-phenanthroline

    Specifications

    Formula Weight 212.21
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    CAS 3922-40-5
    Melting Point >300°C (dec.)
    Sensitivity Hygroscopic
    Physical Form Powder
    MDL Number MFCD00004975
    Synonym 1,10-Phenanthroline-4,7-diol
    Health Hazard 1 Warning
    Color Yellow
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Quantity 0.5g
    Chemical Name or Material 4,7-Dihydroxy-1,10-phenanthroline

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit