Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,8-Dihydroxyquinoline-2-carboxylic Acid 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
59-00-7
C10H7NO4
205.17
MFCD00006754
FBZONXHGGPHHIY-UHFFFAOYSA-N
xanthurenic acid,4,8-dihydroxyquinoline-2-carboxylic acid,xanthuric acid,8-hydroxykynurenic acid,4,8-dihydroxyquinaldic acid,4,8-dihydroxyquinaldinic acid,xanthurenate,2-quinolinecarboxylic acid, 4,8-dihydroxy,unii-58lab1bg8j,ccris 4429
5699
CHEBI:10072
8-hydroxy-4-oxo-1H-quinoline-2-carboxylic acid
C1=CC2=C(C(=C1)O)NC(=CC2=O)C(=O)O
59-00-7
C10H7NO4
205.17
MFCD00006754
FBZONXHGGPHHIY-UHFFFAOYSA-N
xanthurenic acid,4,8-dihydroxyquinoline-2-carboxylic acid,xanthuric acid,8-hydroxykynurenic acid,4,8-dihydroxyquinaldic acid,4,8-dihydroxyquinaldinic acid,xanthurenate,2-quinolinecarboxylic acid, 4,8-dihydroxy,unii-58lab1bg8j,ccris 4429
5699
CHEBI:10072
8-hydroxy-4-oxo-1H-quinoline-2-carboxylic acid
C1=CC2=C(C(=C1)O)NC(=CC2=O)C(=O)O
Melting Point 297°C to 298°C
Color Ochre
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Merck Index 15, 10266
Formula Weight 205.17
Physical Form Powder
Percent Purity ≥95%
Chemical Name or Material 4, 8-Dihydroxyquinoline-2-carboxylic acid, 96%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 200-410-1

RTECSNumber : UZ9275000