Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidobenzenesulfonyl azide, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
2158-14-7
C8H8N4O3S
240.24
NTMHWRHEGDRTPD-UHFFFAOYSA-N
4-acetamidobenzenesulfonyl azide,p-absa,n-4-azidosulfonylphenyl acetamide,4-acetamidobenzenesulfonylazide,4-acetamidobenzenesulphonyl azide,4-acetamidobenzene sulfonyl azide,4-acetamidobenzene-1-sulfonyl azide,acmc-1chyo,ksc491c8r,p-acetamidobenzensulfonyl azide
5129185
N-(4-azidosulfonylphenyl)acetamide
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]
2158-14-7
C8H8N4O3S
240.24
NTMHWRHEGDRTPD-UHFFFAOYSA-N
4-acetamidobenzenesulfonyl azide,p-absa,n-4-azidosulfonylphenyl acetamide,4-acetamidobenzenesulfonylazide,4-acetamidobenzenesulphonyl azide,4-acetamidobenzene sulfonyl azide,4-acetamidobenzene-1-sulfonyl azide,acmc-1chyo,ksc491c8r,p-acetamidobenzensulfonyl azide
5129185
N-(4-azidosulfonylphenyl)acetamide
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]
Infrared Spectrum Authentic
Quantity 25g
Melting Point 107.0°C to 111.0°C
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 240.24
Percent Purity 97%
Chemical Name or Material 4-Acetamidobenzenesulfonyl azide

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning