Alfa Aesar™ 4-Acetamidobenzenesulfonyl chloride, 98+%

Manufacturer: Alfa Aesar™ A115110B

Quantity 1000g

 View more versions of this product

Catalog No. AAA115110B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 121-60-8
    Molecular Formula C8H8ClNO3S
    Formula Weight 233.67
    MDL Number MFCD00007442
    UN Number UN3261
    Beilstein 746676
    Merck Index 14,103
    Synonym N-Acetylsulfanilyl chloride
    Chemical Name or Material 4-Acetamidobenzenesulfonyl chloride

    Specifications

    Formula Weight 233.67
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    TSCA TSCA
    Molecular Formula C8H8ClNO3S
    MDL Number MFCD00007442
    CAS 121-60-8
    UN Number UN3261
    Beilstein 746676
    Merck Index 14,103
    Quantity 1000g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Synonym N-Acetylsulfanilyl chloride
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Melting Point ≈146°C (dec.)
    RTECS Number DB8837500
    Chemical Name or Material 4-Acetamidobenzenesulfonyl chloride
    Assay ≥98%

    Description

    Applications
    N-Acetylsulfanilyl chloride is used in the preparation of sulfanilamide and its derivatives which are intermediates to produce sulfa drugs. They are used in the prevention and treatment of bacterial infections, diabetes mellitus, edema, hypertension, and gout. It is also used as a Pharma raw material. N-Acetylsulfanilyl chloride widely used in the fields of dye, medicine, mainly used for preparation of Sulfanilamide, Sulfanilylureal, Sulfatolamide, Sulphathiourea , Sulfaguanidine and Sulfacetamide and etc.

    Solubility
    It is slightly in water. But soluble in benzene, chloroform and ether.

    Notes
    Stable under ordinary conditions. Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents, Strong bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit