Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Acetamidobenzenesulfonyl chloride, 98+%

Manufacturer:  Alfa Aesar™ A1151130

Quantity 250g

 View more versions of this product

Catalog No. AAA1151130


  Check Availability
Add to cart

Specifications

Description

Applications
N-Acetylsulfanilyl chloride is used in the preparation of sulfanilamide and its derivatives which are intermediates to produce sulfa drugs. They are used in the prevention and treatment of bacterial infections, diabetes mellitus, edema, hypertension, and gout. It is also used as a Pharma raw material. N-Acetylsulfanilyl chloride widely used in the fields of dye, medicine, mainly used for preparation of Sulfanilamide, Sulfanilylureal, Sulfatolamide, Sulphathiourea , Sulfaguanidine and Sulfacetamide and etc.

Solubility
It is slightly in water. But soluble in benzene, chloroform and ether.

Notes
Stable under ordinary conditions. Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents, Strong bases.
Description & Specifications

Chemical Identifiers

121-60-8
233.67
UN3261
14,103
4-Acetamidobenzenesulfonyl chloride
C8H8ClNO3S
MFCD00007442
746676
N-Acetylsulfanilyl chloride

Specifications

121-60-8
233.67
UN3261
14,103
≈146°C (dec.)
4-Acetamidobenzenesulfonyl chloride
C8H8ClNO3S
MFCD00007442
746676
N-Acetylsulfanilyl chloride
250g
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : DB8837500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit