Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidobenzenesulfonyl Chloride 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
121-60-8
C8H8ClNO3S
233.666
MFCD00007442
GRDXCFKBQWDAJH-UHFFFAOYSA-N
n-acetylsulfanilyl chloride,4-acetamidobenzene-1-sulfonyl chloride,dagenan chloride,4-acetylamino benzenesulfonyl chloride,4-acetamidophenylsulfonyl chloride,benzenesulfonyl chloride, 4-acetylamino,acetylsulfanilyl chloride,p-acetamidobenzenesulfonyl chloride,n-acetylsulphanilyl chloride,p-acetaminobenzenesulfonyl chloride
8481
4-acetamidobenzenesulfonyl chloride
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)Cl
121-60-8
C8H8ClNO3S
233.666
MFCD00007442
GRDXCFKBQWDAJH-UHFFFAOYSA-N
n-acetylsulfanilyl chloride,4-acetamidobenzene-1-sulfonyl chloride,dagenan chloride,4-acetylamino benzenesulfonyl chloride,4-acetamidophenylsulfonyl chloride,benzenesulfonyl chloride, 4-acetylamino,acetylsulfanilyl chloride,p-acetamidobenzenesulfonyl chloride,n-acetylsulphanilyl chloride,p-acetaminobenzenesulfonyl chloride
8481
4-acetamidobenzenesulfonyl chloride
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)Cl
Color White-Yellow
Melting Point 146°C
Quantity 100g
UN Number 3261
Formula Weight 233.67
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 4-Acetamidobenzenesulfonyl Chloride

EINECSNumber : (3)-1988

RTECSNumber : DB8837500

TSCA : Yes