Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidophenylboronic acid, 97+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
101251-09-6
C8H10BNO3
178.98
MFCD02179451
VYEWTHXZHHATTA-UHFFFAOYSA-N
4-acetylaminophenylboronic acid,4-acetamidophenyl boronic acid,p-acetamidophenylboronic acid,4-acetamidobenzeneboronic acid,4-acetamidophenyl-boronic acid,4-acetaminophenyl boronic acid,4-acetylaminophenyl boronic acid,4-acetylamino phenylboronic acid,boronic acid, 4-acetylamino phenyl
2734657
(4-acetamidophenyl)boronic acid
B(C1=CC=C(C=C1)NC(=O)C)(O)O
101251-09-6
C8H10BNO3
178.98
MFCD02179451
VYEWTHXZHHATTA-UHFFFAOYSA-N
4-acetylaminophenylboronic acid,4-acetamidophenyl boronic acid,p-acetamidophenylboronic acid,4-acetamidobenzeneboronic acid,4-acetamidophenyl-boronic acid,4-acetaminophenyl boronic acid,4-acetylaminophenyl boronic acid,4-acetylamino phenylboronic acid,boronic acid, 4-acetylamino phenyl
2734657
(4-acetamidophenyl)boronic acid
B(C1=CC=C(C=C1)NC(=O)C)(O)O
Infrared Spectrum Authentic
Quantity 1g
Melting Point 215.0°C to 220.0°C
Packaging Glass bottle
Color Gray-Tan
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97% min. (GC)
Solubility Information Solubility in water: 85g/L (70°C). Other solubilities: slightly soluble in ethylacetate,soluble in meoh,etoh,dmso
Formula Weight 178.98
Physical Form Powder
Percent Purity 97+%
Chemical Name or Material 4-Acetamidophenylboronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C