Alfa Aesar™ 4-Acetoxy-2,5-dimethyl-3(2H)-furanone, 98%

Manufacturer: Alfa Aesar™ B2207909

Quantity 10g

 View more versions of this product

Catalog No. AAB2207909

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 4166-20-5
    Molecular Formula C8H10O4
    Formula Weight 170.17
    MDL Number MFCD00799462
    Synonym Furaneol acetate
    Chemical Name or Material 4-Acetoxy-2,5-dimethyl-3(2H)-furanone

    Specifications

    Formula Weight 170.17
    Molecular Formula C8H10O4
    MDL Number MFCD00799462
    CAS 4166-20-5
    Synonym Furaneol acetate
    Boiling Point 96° to 98°C (1mmHg)
    Quantity 10g
    Chemical Name or Material 4-Acetoxy-2,5-dimethyl-3(2H)-furanone
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit