Alfa Aesar™ 4-Acetoxy-3-ethoxybenzaldehyde, 99%

Manufacturer: Alfa Aesar™ H50488MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50488MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 72207-94-4
    Molecular Formula C11H12O4
    Formula Weight 208.22
    MDL Number MFCD01666413
    Synonym 2-Ethoxy-4-formylphenyl acetate
    Chemical Name or Material 4-Acetoxy-3-ethoxybenzaldehyde

    Specifications

    Formula Weight 208.22
    Molecular Formula C11H12O4
    Quantity 250mg
    RTECS Number CU4381000
    TSCA TSCA
    MDL Number MFCD01666413
    CAS 72207-94-4
    Synonym 2-Ethoxy-4-formylphenyl acetate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4-Acetoxy-3-ethoxybenzaldehyde
    Assay 99%

    Description

    Applications
    4-Acetoxy-3-ethoxybenzaldehyde is used in organic synthesis. It is used as catalytic agent; petrochemical additive.

    Solubility
    Soluble in water (986.4 mg / L) at 25°C.

    Notes
    Air Sensitive. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit