Alfa Aesar™ 4'-Acetoxyacetophenone, 99%

Manufacturer: Alfa Aesar™ B2228436

Quantity 500g

 View more versions of this product

Catalog No. AAB2228436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13031-43-1
    Molecular Formula C10H10O3
    Formula Weight 178.19
    MDL Number MFCD00017229
    Beilstein 2093080
    Synonym 4-Acetylphenyl acetate
    Chemical Name or Material 4'-Acetoxyacetophenone

    Specifications

    Formula Weight 178.19
    TSCA TSCA
    Molecular Formula C10H10O3
    MDL Number MFCD00017229
    CAS 13031-43-1
    Beilstein 2093080
    Synonym 4-Acetylphenyl acetate
    Boiling Point 165° to 170°C (14mmHg)
    Melting Point 51° to 54°C
    Quantity 500g
    Chemical Name or Material 4'-Acetoxyacetophenone
    Assay 99%

    Description

    Applications
    4 ft.-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.

    Solubility
    Slightly soluble in water(4450 mg/L).

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit