Alfa Aesar™ 4-Acetoxybiphenyl, 98%

Manufacturer: Alfa Aesar™ A1216618

Quantity 50g

 View more versions of this product

Catalog No. AAA1216618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 148-86-7
    Molecular Formula C14H12O2
    Formula Weight 212.25
    MDL Number MFCD00014979
    Beilstein 1872019
    Synonym 4-Biphenylyl acetate
    Chemical Name or Material 4-Acetoxybiphenyl

    Specifications

    Formula Weight 212.25
    MDL Number MFCD00014979
    CAS 148-86-7
    Molecular Formula C14H12O2
    Beilstein 1872019
    Synonym 4-Biphenylyl acetate
    Boiling Point 196°C
    Melting Point 85° to 90°C
    Quantity 50g
    Chemical Name or Material 4-Acetoxybiphenyl
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit