Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Acetyl-4-methylpimelic acid, 97%

Click to view available options
Quantity:
1g
5g
19830-09-2
C10H16O5
216.233
MFCD00052871
LYBQMKJABQTTFJ-UHFFFAOYSA-N
4-acetyl-4-methylpimelic acid,aav2ytm2mi,unii-aav2ytm2mi,3-acetyl-3-methylpentane-1,5-dicarboxylic acid,acmc-1c6z0,4-acetyl-4-methyl-heptanedioic acid,heptanedioic acid,4-acetyl-4-methyl
407651
4-acetyl-4-methylheptanedioic acid
CC(=O)C(C)(CCC(=O)O)CCC(=O)O
19830-09-2
C10H16O5
216.233
MFCD00052871
LYBQMKJABQTTFJ-UHFFFAOYSA-N
4-acetyl-4-methylpimelic acid,aav2ytm2mi,unii-aav2ytm2mi,3-acetyl-3-methylpentane-1,5-dicarboxylic acid,acmc-1c6z0,4-acetyl-4-methyl-heptanedioic acid,heptanedioic acid,4-acetyl-4-methyl
407651
4-acetyl-4-methylheptanedioic acid
CC(=O)C(C)(CCC(=O)O)CCC(=O)O
Melting Point 121°C to 125°C
Quantity 5g
Beilstein 1789115
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material 4-Acetyl-4-methylpimelic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures