Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Acetylbenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
149104-90-5
C8H9BO3
163.967
MFCD01074667
OBQRODBYVNIZJU-UHFFFAOYSA-N
4-acetylphenyl boronic acid,p-acetylphenylboronic acid,4-acetylbenzeneboronic acid,1-4-dihydroxyboranyl phenyl ethan-1-one,boronic acid, 4-acetylphenyl,4acetylphenylboronic acid,4-actylphenylboronic acid,4acetylbenzeneboronic acid,p-acetylbenzeneboronic acid
3702122
(4-acetylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)C)(O)O
149104-90-5
C8H9BO3
163.967
MFCD01074667
OBQRODBYVNIZJU-UHFFFAOYSA-N
4-acetylphenyl boronic acid,p-acetylphenylboronic acid,4-acetylbenzeneboronic acid,1-4-dihydroxyboranyl phenyl ethan-1-one,boronic acid, 4-acetylphenyl,4acetylphenylboronic acid,4-actylphenylboronic acid,4acetylbenzeneboronic acid,p-acetylbenzeneboronic acid
3702122
(4-acetylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)C)(O)O
Melting Point ~240°C (decomposition)
Quantity 1g
Beilstein 7869162
Formula Weight 163.97
Percent Purity 98%
Chemical Name or Material 4-Acetylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning