Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylbenzenesulfonyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
50g
Packaging:
Glass bottle
1788-10-9
C8H7ClO3S
218.65
MFCD00800269
FXVDNCRTKXMSEZ-UHFFFAOYSA-N
4-acetylbenzene-1-sulfonyl chloride,4-acetyl-benzenesulfonyl chloride,4-acetylphenylsulfonyl chloride,benzenesulfonylchloride, 4-acetyl,benzenesulfonyl chloride, 4-acetyl,1-acetyl-4-chlorosulfonyl benzene,4-acetylbenzenesulfonyl chloride,acmc-1bs75,ksc183k3b,4-acetylbenzenesulfonylchloride
266418
4-acetylbenzenesulfonyl chloride
CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl
1788-10-9
C8H7ClO3S
218.65
MFCD00800269
FXVDNCRTKXMSEZ-UHFFFAOYSA-N
4-acetylbenzene-1-sulfonyl chloride,4-acetyl-benzenesulfonyl chloride,4-acetylphenylsulfonyl chloride,benzenesulfonylchloride, 4-acetyl,benzenesulfonyl chloride, 4-acetyl,1-acetyl-4-chlorosulfonyl benzene,4-acetylbenzenesulfonyl chloride,acmc-1bs75,ksc183k3b,4-acetylbenzenesulfonylchloride
266418
4-acetylbenzenesulfonyl chloride
CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl
Color Brown to White
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Melting Point 83°C to 87°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula H3CCO(C6H4)SO2Cl
Formula Weight 218.65
Physical Form Powder and/or Chunks
Percent Purity 97%
Chemical Name or Material 4-Acetylbenzenesulfonyl chloride, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger