Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Acetylbenzoic acid, 98+%

Click to view available options
Quantity:
1g
5g
25g
586-89-0
C9H8O3
164.16
MFCD00002561
QBHDSQZASIBAAI-UHFFFAOYSA-N
benzoic acid, 4-acetyl,p-acetylbenzoic acid,4-acetyl-benzoic acid,4'-acetophenonecarboxylic acid,4-acetylbezoic acid,4-acetylbenzoci acid,4'-carboxyacetohenone,pubchem3129,4-acetyl benzoic acid
11470
4-acetylbenzoic acid
CC(=O)C1=CC=C(C=C1)C(=O)O
586-89-0
C9H8O3
164.16
MFCD00002561
QBHDSQZASIBAAI-UHFFFAOYSA-N
benzoic acid, 4-acetyl,p-acetylbenzoic acid,4-acetyl-benzoic acid,4'-acetophenonecarboxylic acid,4-acetylbezoic acid,4-acetylbenzoci acid,4'-carboxyacetohenone,pubchem3129,4-acetyl benzoic acid
11470
4-acetylbenzoic acid
CC(=O)C1=CC=C(C=C1)C(=O)O
Applications
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Quantity 5g
Odor Odorless
Melting Point 208°C to 212°C
Beilstein 2207355
Solubility Information Soluble in water.
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material 4-Acetylbenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-588-5

TSCA : No

Recommended Storage : Ambient temperatures