Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
149104-90-5
C8H9BO3
163.97
MFCD01074667
OBQRODBYVNIZJU-UHFFFAOYSA-N
4-acetylphenyl boronic acid,p-acetylphenylboronic acid,4-acetylbenzeneboronic acid,1-4-dihydroxyboranyl phenyl ethan-1-one,boronic acid, 4-acetylphenyl,4acetylphenylboronic acid,4-actylphenylboronic acid,4acetylbenzeneboronic acid,p-acetylbenzeneboronic acid
3702122
(4-acetylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)C)(O)O
149104-90-5
C8H9BO3
163.97
MFCD01074667
OBQRODBYVNIZJU-UHFFFAOYSA-N
4-acetylphenyl boronic acid,p-acetylphenylboronic acid,4-acetylbenzeneboronic acid,1-4-dihydroxyboranyl phenyl ethan-1-one,boronic acid, 4-acetylphenyl,4acetylphenylboronic acid,4-actylphenylboronic acid,4acetylbenzeneboronic acid,p-acetylbenzeneboronic acid
3702122
(4-acetylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)C)(O)O
Infrared Spectrum Authentic
Color Yellow
Quantity 25g
Melting Point 243.0°C to 247.0°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 163.97
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Acetylphenylboronic acid
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning