Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Acetylpyridazine, 95%

Click to view available options
Quantity:
250mg
1g
50901-46-7
C6H6N2O
122.127
MFCD13193354
IWFYWGOLMZIBQB-UHFFFAOYSA-N
1-pyridazin-4-yl ethanone,1-pyridazin-4-yl ethan-1-one,4-acetylpyridazine,ethanone, 1-4-pyridazinyl,4-acetyl-pyridazin,1-pyridazin-4-yl-ethanone,methyl 4-pyridazinyl ketone,1-pyridazin-4-yl ;ethanone,1-4-pyridazinyl-ethanone,ethanone,1-4-pyridazinyl-9ci
14452005
1-pyridazin-4-ylethanone
CC(=O)C1=CN=NC=C1
50901-46-7
C6H6N2O
122.127
MFCD13193354
IWFYWGOLMZIBQB-UHFFFAOYSA-N
1-pyridazin-4-yl ethanone,1-pyridazin-4-yl ethan-1-one,4-acetylpyridazine,ethanone, 1-4-pyridazinyl,4-acetyl-pyridazin,1-pyridazin-4-yl-ethanone,methyl 4-pyridazinyl ketone,1-pyridazin-4-yl ;ethanone,1-4-pyridazinyl-ethanone,ethanone,1-4-pyridazinyl-9ci
14452005
1-pyridazin-4-ylethanone
CC(=O)C1=CN=NC=C1
Quantity 250mg
Beilstein 742138
Formula Weight 122.13
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4-Acetylpyridazine

storageNote1 : -30° to -10°C