Learn More
4-Acetylpyridine, 98%, Thermo Scientific Chemicals
Description
4-acetylpyridine on reaction with - and 4-formylpyridine (H3FoPyS, H4FoPyS) yields semicarbazones. Reaction of acetylpyridine with thiophene-2-carboxaldehyde afforded the 2-chloro-6-ethoxy-4-β-(2-thienyl)acryloylpyridine, which was reacted with malononitrile in refluxing ethanol in the presence of piperidine as a catalyst to afford the cyanoaminopyrane derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-acetylpyridine on reaction with - and 4-formylpyridine (H3FoPyS, H4FoPyS) yields semicarbazones. Reaction of acetylpyridine with thiophene-2-carboxaldehyde afforded the 2-chloro-6-ethoxy-4-β-(2-thienyl)acryloylpyridine, which was reacted with malononitrile in refluxing ethanol in the presence of piperidine as a catalyst to afford the cyanoaminopyrane derivative.
Solubility
Insoluble in water. Soluble in alcohol.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
Melting Point | 14°C to 16°C |
Density | 1.103 |
Boiling Point | 210°C to 212°C |
Flash Point | 104°C (219°F) |
Refractive Index | 1.535 |
Quantity | 100 g |
Beilstein | 107629 |
Solubility Information | Insoluble in water. Soluble in alcohol. |
Formula Weight | 121.14 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 214-350-9 |
RTECSNumber | OB5426000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.