Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Acetyltetrahydropyran, 97%

Click to view available options
Quantity:
250mg
1g
137052-08-5
C7H12O2
128.171
MFCD08704647
VNMXIOWPBADSIC-UHFFFAOYSA-N
1-tetrahydro-2h-pyran-4-yl ethanone,1-oxan-4-yl ethan-1-one,1-oxan-4-yl ethanone,4-acetyltetrahydropyran,1-tetrahydro-2h-pyran-4-ylethanone,4-acetyltetrahydro-4h-pyran,4-acetyltetrahydro-2h-pyran,1-tetrahydropyran-4-ylethanone,1-tetrahydro-pyran-4-yl-ethanone,1-tetrahydro-2h-pyran-4-yl ethan-1-one
9877365
1-(oxan-4-yl)ethanone
CC(=O)C1CCOCC1
137052-08-5
C7H12O2
128.171
MFCD08704647
VNMXIOWPBADSIC-UHFFFAOYSA-N
1-tetrahydro-2h-pyran-4-yl ethanone,1-oxan-4-yl ethan-1-one,1-oxan-4-yl ethanone,4-acetyltetrahydropyran,1-tetrahydro-2h-pyran-4-ylethanone,4-acetyltetrahydro-4h-pyran,4-acetyltetrahydro-2h-pyran,1-tetrahydropyran-4-ylethanone,1-tetrahydro-pyran-4-yl-ethanone,1-tetrahydro-2h-pyran-4-yl ethan-1-one
9877365
1-(oxan-4-yl)ethanone
CC(=O)C1CCOCC1
Refractive Index 1.4530
Boiling Point 90°C to 94°C (15mmHg)
Density 1.024g/mL
Quantity 250mg
Color Colorless to Yellow
Beilstein 109587
Formula Weight 128.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Acetyltetrahydropyran

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning