Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acryloylmorpholine, 97%, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
Packaging:
Glass bottle
5117-12-4
C7H11NO2
141.17
XLPJNCYCZORXHG-UHFFFAOYSA-N
methyl 3-hydroxy-2-thiophenecarboxylate,3-hydroxy-2-thiophenecarboxylic acid methyl ester,3-hydroxy-thiophene-2-carboxylic acid methyl ester,3-hydroxythiophene-2-carboxylic acid methyl ester,pubchem7744,acmc-1aqhj,maybridge3_005989,3-hydroxy-2-methoxycarbonylthiophene,3-hydroxy-2-methoxycarbonyl-thiophene,methyl 3-hydroxythiophen-2-caboxylate
581127
methyl 3-hydroxythiophene-2-carboxylate
C=CC(=O)N1CCOCC1
5117-12-4
C7H11NO2
141.17
XLPJNCYCZORXHG-UHFFFAOYSA-N
methyl 3-hydroxy-2-thiophenecarboxylate,3-hydroxy-2-thiophenecarboxylic acid methyl ester,3-hydroxy-thiophene-2-carboxylic acid methyl ester,3-hydroxythiophene-2-carboxylic acid methyl ester,pubchem7744,acmc-1aqhj,maybridge3_005989,3-hydroxy-2-methoxycarbonylthiophene,3-hydroxy-2-methoxycarbonyl-thiophene,methyl 3-hydroxythiophen-2-caboxylate
581127
methyl 3-hydroxythiophene-2-carboxylate
C=CC(=O)N1CCOCC1
Water 2.5% Max.
Specific Gravity 1.122
Density 1.1220g/mL
Flash Point 113°C
Quantity 250mL
Packaging Glass bottle
Boiling Point 158.0°C (50.0 mmHg)
Color Colorless to Yellow
Infrared Spectrum Authentic
Melting Point -35.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 141.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Acryloylmorpholine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water.
Get medical attention/advice if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger