Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Allylmorpholine 97.0+%, TCI America™

Click to view available options
Quantity:
5mL
25mL
696-57-1
C7H13NO
127.187
MFCD00038034
SUSANAYXICMXBL-UHFFFAOYSA-N
4-allylmorpholine,n-allylmorpholine,allylmorpholine,n-allyl morpholine,4-prop-2-en-1-yl morpholine,allyl morpholine,n-allymorpholine,4-allyl-morpholine,4-allylmorpholine #,morpholine, 4-allyl
136522
4-prop-2-enylmorpholine
C=CCN1CCOCC1
696-57-1
C7H13NO
127.187
MFCD00038034
SUSANAYXICMXBL-UHFFFAOYSA-N
4-allylmorpholine,n-allylmorpholine,allylmorpholine,n-allyl morpholine,4-prop-2-en-1-yl morpholine,allyl morpholine,n-allymorpholine,4-allyl-morpholine,4-allylmorpholine #,morpholine, 4-allyl
136522
4-prop-2-enylmorpholine
C=CCN1CCOCC1
Boiling Point 158°C
Color Yellow
Quantity 25mL
UN Number 1993
Formula Weight 127.19
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material 4-Allylmorpholine