Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Allyloxybenzoyl chloride, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
36844-51-6
C10H9ClO2
196.63
MFCD09744024
WYMVIVUKHYHNBN-UHFFFAOYSA-N
4-allyloxybenzoyl chloride,benzoyl chloride, 4-2-propen-1-yloxy,4-prop-2-en-1-yloxy benzoyl chloride,p-allyloxybenzoyl chloride,4-allyloxy benzoyl chloride,4-2-propenyloxy benzoyl chloride,4-allyloxy benzoic acid chloride,4-prop-2-en-1-oxy benzoyl chloride,benzoyl chloride,4-2-propen-1-yloxy
11790137
4-prop-2-enoxybenzoyl chloride
C=CCOC1=CC=C(C=C1)C(=O)Cl
36844-51-6
C10H9ClO2
196.63
MFCD09744024
WYMVIVUKHYHNBN-UHFFFAOYSA-N
4-allyloxybenzoyl chloride,benzoyl chloride, 4-2-propen-1-yloxy,4-prop-2-en-1-yloxy benzoyl chloride,p-allyloxybenzoyl chloride,4-allyloxy benzoyl chloride,4-2-propenyloxy benzoyl chloride,4-allyloxy benzoic acid chloride,4-prop-2-en-1-oxy benzoyl chloride,benzoyl chloride,4-2-propen-1-yloxy
11790137
4-prop-2-enoxybenzoyl chloride
C=CCOC1=CC=C(C=C1)C(=O)Cl
Specific Gravity 1.155
Color Undesignated
Density 1.1550g/mL
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula CH2CHCH2OC6H4COCl
Formula Weight 196.63
Physical Form Low Melting Solid
Percent Purity 98%
Chemical Name or Material 4-Allyloxybenzoyl chloride
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger