Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1,2,2,6,6-pentamethylpiperidine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
40327-96-6
C10H22N2
170.3
MFCD01861832
CGXOAAMIQPDTPE-UHFFFAOYSA-N
4-amino-1,2,2,6,6-pentamethylpiperidine,1,2,2,6,6-pentamethyl-4-aminopiperidine,1,2,2,6,6-pentamethyl-4-piperidinamine,1,2,2,6,6-pentamethyl-4-piperidylamine,4-ammo-1,2,2,6,6-pentamethylpiperidine,4-piperidinamine,1,2,2,6,6-pentamethyl,1,2,-2,6,6-pentamethyl-4-aminopiperidine,1,2,2,6,6,-pentamethyl-4-aminopiperidine,4-amino-1,2,2,6,6-pentamethyl-piperidine,4-amino-1,2,2,6,6-pentamethyl piperidine, 97+%
693792
1,2,2,6,6-pentamethylpiperidin-4-amine
CC1(CC(CC(N1C)(C)C)N)C
40327-96-6
C10H22N2
170.3
MFCD01861832
CGXOAAMIQPDTPE-UHFFFAOYSA-N
4-amino-1,2,2,6,6-pentamethylpiperidine,1,2,2,6,6-pentamethyl-4-aminopiperidine,1,2,2,6,6-pentamethyl-4-piperidinamine,1,2,2,6,6-pentamethyl-4-piperidylamine,4-ammo-1,2,2,6,6-pentamethylpiperidine,4-piperidinamine,1,2,2,6,6-pentamethyl,1,2,-2,6,6-pentamethyl-4-aminopiperidine,1,2,2,6,6,-pentamethyl-4-aminopiperidine,4-amino-1,2,2,6,6-pentamethyl-piperidine,4-amino-1,2,2,6,6-pentamethyl piperidine, 97+%
693792
1,2,2,6,6-pentamethylpiperidin-4-amine
CC1(CC(CC(N1C)(C)C)N)C
Boiling Point 215°C to 218°C
Flash Point 72°C
Refractive Index 1.4795 to 1.4815
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 5g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 22, II, 321
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Formula Weight 170.3
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 4-Amino-1,2,2,6,6-pentamethylpiperidine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger